2-Amino-5-chlorobenzoxazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Chlorobenzo[d]oxazol-2-amine, Zoxazolamine, 2-Amino-5-chloro-1,3-benzoxazole
M-Clarity™ quality level = MQ100
A45807-1G A45807-25G A45807-5G
SIALA45807-1GEA 44.6 EUR
SIALA45807-1G SIALA45807-25G SIALA45807-5G
2-Amino-5-chlorobenzoxazole, Sigma-Aldrich®
2-Amino-5-chlorobenzoxazole
Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 184 °C
MDL Number: MFCD00005770
CAS Number: 61-80-3
EINECS: 200-519-4
Merck Index: 12,10328