5-Amino-1H-indazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine
M-Clarity™ quality level = MQ100
A59557-25G A59557-5G
SIALA59557-25GEA 227 EUR
SIALA59557-25G SIALA59557-5G
5-Amino-1H-indazole, Sigma-Aldrich®
5-Amino-1H-indazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 183 °C
MDL Number: MFCD00037975
CAS Number: 19335-11-6
EINECS: 242-971-5
UN: 2811
ADR: 6.1,III