11-Aminoundecanoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
11-Aminoundecylic acid
M-Clarity™ quality level = MQ200
A82605-100G
A82605-500G
SIALA82605-100GEA
61.2
EUR
SIALA82605-100G
SIALA82605-500G
11-Aminoundecanoic acid, Sigma-Aldrich®
11-Aminoundecanoic acid
Formula:
C₁₁H₂₃NO₂ Boiling Pt: 331 °C (1013 hPa) Melting Pt: 187…190 °C |
MDL Number:
MFCD00008150 CAS Number: 2432-99-7 EINECS: 219-417-6 |