7-Azaindole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1H-Pyrrolo[2,3-b]pyridine
M-Clarity™ quality level = MQ100
A95502-1G A95502-5G
SIALA95502-1GEA 36.8 EUR
SIALA95502-1G SIALA95502-5G
7-Azaindole, Sigma-Aldrich®
7-Azaindole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 103…106 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0