3-Benzoylpropionic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid
M-Clarity™ quality level = MQ100
B13802-100G
B13802-25G
SIALB13802-100GEA
289
EUR
SIALB13802-100G
SIALB13802-25G
3-Benzoylpropionic acid, Sigma-Aldrich®
3-Benzoylpropionic acid
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol Boiling Pt: 325 °C (1013 hPa) Melting Pt: 114…117 °C |
MDL Number:
MFCD00002792 CAS Number: 2051-95-8 EINECS: 218-135-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...