4-Phenylbenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid
M-Clarity™ quality level = MQ100
B34729-100G B34729-25G
SIALB34729-100GEA 94 EUR
SIALB34729-100G SIALB34729-25G
4-Phenylbenzoic acid, Sigma-Aldrich®
4-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
Boiling Pt: 373 °C (1013 hPa)
Melting Pt: 220 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002553
CAS Number: 92-92-2
EINECS: 202-203-1