Benzofuran, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone
M-Clarity™ quality level = MQ100
B8002-25G B8002-5G
SIALB8002-25GEA 64.9 EUR
SIALB8002-25G SIALB8002-5G
Benzofuran, Sigma-Aldrich®
Benzofuran
Formula: C₈H₆O
MW: 118.14 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: < –18 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00005847
CAS Number: 271-89-6
EINECS: 205-982-6
Merck Index: 13,01089