(4-Chlorophenoxy)acetic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4-CPA
M-Clarity™ quality level = MQ200
C0413-100G
C0413-25G
C0413-500G
SIALC0413-100GEA
106
EUR
SIALC0413-100G
SIALC0413-25G
SIALC0413-500G
(4-Chlorophenoxy)acetic acid, Sigma-Aldrich®
(4-Chlorophenoxy)acetic acid
Formula:
(C₆H₄Cl)OCH₂COOH MW: 186.59 g/mol Boiling Pt: 308 °C (1013 hPa) Melting Pt: 160 °C Density: 1.53 g/cm³ (20 °C) |
MDL Number:
MFCD00004305 CAS Number: 122-88-3 EINECS: 204-581-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...