Bis(2-chloroethyl)ether, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Chloroethyl ether
M-Clarity™ quality level = MQ200
C41134-100G C41134-500G
SIALC41134-100GEA 67.8 EUR
SIALC41134-100G SIALC41134-500G
Bis(2-chloroethyl)ether, Sigma-Aldrich®
Bis(2-chloroethyl) ether
Formula: C₄H₈Cl₂O
MW: 143.01 g/mol
Boiling Pt: 178
Melting Pt: –44 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 55
MDL Number: MFCD00000975
CAS Number: 111-44-4
EINECS: 203-870-1
UN: 1916
ADR: 6.1 II
Merck Index: 13,03091