trans-Cinnamaldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C80687-1KG C80687-500G C80687-25G
SIALC80687-1KGEA 72.9 EUR
SIALC80687-1KG SIALC80687-500G SIALC80687-25G
trans-Cinnamaldehyde, Sigma-Aldrich®
trans-Cinnamaldehyde
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00007000
CAS Number: 14371-10-9
Merck Index: 12,02319