Cyanoacetic acid, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
C88505-1KG C88505-25G C88505-500G
SIALC88505-1KGEA 196 EUR
SIALC88505-1KG SIALC88505-25G SIALC88505-500G
Cyanoacetic acid, Sigma-Aldrich®
Cyanoacetic acid
Formula: C₃H₃NO₂
MW: 85.06 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 66 °C
Density: >1.1 g/cm³ (20 °C)
Flash Pt: >165 °C
MDL Number: MFCD00002677
CAS Number: 372-09-8
EINECS: 206-743-9
UN: 3261
ADR: 8,II
Merck Index: 13,02718