4,5-Dichloro-o-phenylenediamine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine
M-Clarity™ quality level = MQ200
D71607-25G
D71607-5G
SIALD71607-25GEA
306
EUR
SIALD71607-25G
SIALD71607-5G
4,5-Dichloro-o-phenylenediamine, Sigma-Aldrich®
4,5-Dichloro-o-phenylenediamine
Formula:
C₆H₆Cl₂N₂ MW: 177.03 g/mol Boiling Pt: 305 °C (1013 hPa) Melting Pt: 163 °C |
MDL Number:
MFCD00007723 CAS Number: 5348-42-5 EINECS: 226-305-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...