(2,4-Dichlorophenoxy)acetic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,4-D
M-Clarity™ quality level = MQ200
D7299-100G D7299-250G
SIALD7299-100GEA 69.6 EUR
SIALD7299-100G SIALD7299-250G
(2,4-Dichlorophenoxy)acetic acid, Sigma-Aldrich®
(2,4-Dichlorophenoxy)acetic acid
Formula: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04 g/mol
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 137…139 °C
Density: 1.563 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004300
CAS Number: 94-75-7
EINECS: 202-361-1
Merck Index: 13,02825