Ethyl cyanoacetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
E18425-1KG E18425-250G E18425-5G
SIALE18425-1KGEA 117 EUR
SIALE18425-1KG SIALE18425-250G SIALE18425-5G
Ethyl cyanoacetate, Sigma-Aldrich®
Ethyl cyanoacetate
Formula: C₅H₇NO₂
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.063 g/cm³ (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00001940
CAS Number: 105-56-6
EINECS: 203-309-0
Merck Index: 13,03821