2-Ethyl-4-methylimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Ethyl-4-methyl-1H-imidazole
M-Clarity™ quality level = MQ200
E36652-100G E36652-5G
SIALE36652-100GEA 76.3 EUR
SIALE36652-100G SIALE36652-5G
2-Ethyl-4-methylimidazole, Sigma-Aldrich®
2-Ethyl-4-methylimidazole
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 292
Melting Pt: 45
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00005193
CAS Number: 931-36-2
EINECS: 213-234-5