5-Fluorouracil, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
M-Clarity™ quality level = MQ200
F8423-10G F8423-5G
SIALF8423-10GEA 871 EUR
SIALF8423-10G SIALF8423-5G
5-Fluorouracil, Sigma-Aldrich®
5-Fluorouracil
Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 282…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,04208