5-Hexen-2-one, Sigma-Aldrich®

Supplier: Merck

Synonyms: Hex-5-en-2-one, Hex-1-en-5-one, 1-Hexen-5-one

M-Clarity™ quality level = MQ200

H13001-100G H13001-25G H13001-5G
SIALH13001-100GEA 325 EUR
SIALH13001-100G SIALH13001-25G SIALH13001-5G
5-Hexen-2-one, Sigma-Aldrich®
5-Hexen-2-one
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 129 °C (1013 hPa)
Density: 0.843 g/cm³ (20 °C)
MDL Number: MFCD00008793
CAS Number: 109-49-9
EINECS: 203-675-1
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR