9-Hydroxyfluorene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 9-Fluorenol, Fluoren-9-ol
M-Clarity™ quality level = MQ100
H31204-5G H31204-25G
SIALH31204-5GEA 53 EUR
SIALH31204-5G SIALH31204-25G
9-Hydroxyfluorene, Sigma-Aldrich®
9-Hydroxyfluorene
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 152…157 °C
Density: 1.151 g/cm³ (20 °C)
MDL Number: MFCD00001135
CAS Number: 1689-64-1
EINECS: 216-879-0