Indazole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1H-Indazole

M-Clarity™ quality level = MQ100

I2401-1G I2401-5G
SIALI2401-1GEA 39.2 EUR
SIALI2401-1G SIALI2401-5G
Indazole, Sigma-Aldrich®
Indazole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 270
Melting Pt: 147…149 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00005691
CAS Number: 271-44-3
EINECS: 205-978-4
Merck Index: 13,04961

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR