Levulinic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid
M-Clarity™ quality level = MQ200
L2009-1KG L2009-250G L2009-50G
SIALL2009-1KGEA 129 EUR
SIALL2009-1KG SIALL2009-250G SIALL2009-50G
Levulinic acid, Sigma-Aldrich®
Levulinic acid
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 26…29 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00002796
CAS Number: 123-76-2
EINECS: 204-649-2
UN: 3261
ADR: 8,III
Merck Index: 13,05492