4-Methoxybenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALM10204-25GEA 64.6 EUR
SIALM10204-25G SIALM10204-5G
4-Methoxybenzenesulphonyl chloride, Sigma-Aldrich®
4-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206,65 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 40…42 °C
MDL Number: MFCD00007446
UN: 3261
ADR: 8,II