1-Amino-3-methoxypropane, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Methoxypropylamine

M-Clarity™ quality level = MQ100

M25007-1L M25007-25ML
SIALM25007-1LEA 50 EUR
SIALM25007-1L SIALM25007-25ML
1-Amino-3-methoxypropane, Sigma-Aldrich®
1-Amino-3-methoxypropane
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 118
Melting Pt: < –70 °C
Density: 0.8737 g/cm³ (20 °C)
Flash Pt: 27
MDL Number: MFCD00014831
CAS Number: 5332-73-0
EINECS: 226-241-3
UN: 2734
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR