Methyl-2,4-dihydroxybenzoate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,4-Dihydroxybenzoic acid methyl ester, Methyl 2,4-dihydroxybenzoate

M-Clarity™ quality level = MQ100

M42505-100G M42505-25G
SIALM42505-100GEA 316 EUR
SIALM42505-100G SIALM42505-25G
Methyl-2,4-dihydroxybenzoate, Sigma-Aldrich®
Methyl 2,4-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 214…215 °C
MDL Number: MFCD00002276
CAS Number: 2150-47-2
EINECS: 218-428-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR