2-Methylfuran, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
M46845-100ML M46845-500ML M46845-5ML
SIALM46845-100MLEA 48.1 EUR
SIALM46845-100ML SIALM46845-500ML SIALM46845-5ML
2-Methylfuran, Sigma-Aldrich®
2-Methylfuran
Formula: C₅H₆O
MW: 82.1 g/mol
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.927 g/cm³ (20 °C)
MDL Number: MFCD00003248
CAS Number: 534-22-5
EINECS: 208-594-5
UN: 2301
ADR: 3,II