4-(Methylthio)aniline, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-(Methylmercapto)aniline

M-Clarity™ quality level = MQ100

M54503-25G M54503-5G
SIALM54503-25GEA 78.2 EUR
SIALM54503-25G SIALM54503-5G
4-(Methylthio)aniline, Sigma-Aldrich®
4-(Methylthio)aniline
Formula: C₇H₉NS
MW: 139.22 g/mol
Boiling Pt: 74 °C (0.1 mmHg)
Melting Pt: 156…158 °C
Density: 1.1379 g/cm³ (20 °C)
MDL Number: MFCD00007889
CAS Number: 104-96-1
EINECS: 203-256-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR