2,2'-Phenyliminodiethanol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
N-Phenyl-2,2'-iminodiethanol, 2,2-(Phenylimino)diethanol, N-Phenyldiethanolamine
M-Clarity™ quality level = MQ200
P22400-250G
P22400-1KG
SIALP22400-250GEA
35
EUR
SIALP22400-250G
SIALP22400-1KG
2,2'-Phenyliminodiethanol, Sigma-Aldrich®
2,2'-Phenyliminodiethanol
Formula:
(C₆H₅)N(CH₂CH₂OH)₂ MW: 181.23 g/mol Boiling Pt: 348 °C (1013 hPa) Melting Pt: 56…59 °C Density: 1.201 g/cm³ (60 °C) Flash Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002845 CAS Number: 120-07-0 EINECS: 204-368-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...