2-Pyridylmethanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Pyridylmethanol, 2-Pyridylcarbinol, 2-Pyridinemethanol, Pyridin-2-ylmethanol, 2-(Hydroxymethyl)pyridine, Pyridine-2-methanol
M-Clarity™ quality level = MQ200
P66602-100G P66602-25G
SIALP66602-100GEA 65.1 EUR
SIALP66602-100G SIALP66602-25G
2-Pyridylmethanol, Sigma-Aldrich®
2-Pyridylmethanol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.1317 g/cm³ (20 °C)
MDL Number: MFCD00006348
CAS Number: 586-98-1
EINECS: 209-592-7