2-Pyrrolecarbaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1H-Pyrrole-2-carboxaldehyde, Pyrrole-2-carboxaldehyde, 1H-Pyrrole-2-carbaldehyde, Pyrrole-2-carbaldehyde
M-Clarity™ quality level = MQ200
P73404-100G P73404-25G P73404-5G
SIALP73404-100GEA 205 EUR
SIALP73404-100G SIALP73404-25G SIALP73404-5G
2-Pyrrolecarbaldehyde, Sigma-Aldrich®
2-Pyrrolecarbaldehyde
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 43…46 °C
MDL Number: MFCD00005217
CAS Number: 1003-29-8
EINECS: 213-705-5