Tetrasodium diphosphate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Sodium pyrophosphate tetrabasic, Tetrasodium diphosphate, Sodium diphosphate, Sodium pyrophosphate, tetra-Sodium pyrophosphate
P8010-2.5KG
P8010-500G
SIALP8010-2.5KGEA
301
EUR
SIALP8010-2.5KG
SIALP8010-500G
Tetrasodium diphosphate, Sigma-Aldrich®
tetra-Sodium diphosphate
Formula:
Na₄O₇P₂ MW: 265.9 g/mol Melting Pt: 988 Density: 2.534 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003513 CAS Number: 7722-88-5 EINECS: 231-767-1 Merck Index: 13,09312 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...