(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA
M-Clarity™ quality level = MQ100
T12653-2.5L T12653-500ML
SIALT12653-2.5LEA 83.8 EUR
SIALT12653-2.5L SIALT12653-500ML
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
(±)-Tetrahydrofurfuryl alcohol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 178
Melting Pt: < –80 °C
Density: 1.0543 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353