Tetramethylurea, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: N,N,N',N'-Tetramethylurea, 1,1,3,3-Tetramethylurea

MQ200

T24503-100G T24503-5G
SIALT24503-100GEA 95.9 EUR
SIALT24503-100G SIALT24503-5G
Tetramethylurea, Sigma-Aldrich®
Tetramethylurea
Formula: C₅H₁₂N₂O
MW: 116.16 g/mol
Boiling Pt: 175
Melting Pt: –1 °C
Density: 1.165 g/cm³ (20 °C)
Flash Pt: 75
Storage Temperature: Ambient
MDL Number: MFCD00008319
CAS Number: 632-22-4
EINECS: 211-173-9
Merck Index: 12,09368

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR