2-Thiopheneethanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-(2-Thienyl)ethanol, Thiophen-2-ethanol
M-Clarity™ quality level = MQ200
T27855-100G T27855-25G T27855-5G
SIALT27855-100GEA 526 EUR
SIALT27855-100G SIALT27855-25G SIALT27855-5G
2-Thiopheneethanol, Sigma-Aldrich®
2-Thiopheneethanol
Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 108 °C (1013 hPa)
Density: 1.197 g/cm³ (20 °C)
MDL Number: MFCD00005462
CAS Number: 5402-55-1
EINECS: 226-452-0