2-Thenoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Thiophene-2-carboxylic acid, 2-Thiophenecarboxylic acid
M-Clarity™ quality level = MQ100
T32603-100G T32603-25G
SIALT32603-100GEA 60.7 EUR
SIALT32603-100G SIALT32603-25G
2-Thenoic acid, Sigma-Aldrich®
2-Thenoic acid
Formula: C₅H₄O₂S
MW: 128.15 g/mol
Boiling Pt: 260
Melting Pt: 123…125 °C
MDL Number: MFCD00005437
CAS Number: 527-72-0
EINECS: 208-423-4
Merck Index: 12,09491