3,3',5,5'-Tetramethylbenzidine (TMB) substrate 1 mg, tablets, Sigma-Aldrich®
Supplier: Merck
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
M-Clarity™ quality level = MQ200
T5525-100TAB T5525-50TAB
SIALT5525-100TABEA 583 EUR
SIALT5525-100TAB SIALT5525-50TAB
3,3',5,5'-Tetramethylbenzidine (TMB) substrate 1 mg, tablets, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidine (TMB)
Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00007748
CAS Number: 54827-17-7
EINECS: 259-364-6