Trimethylsulphonium iodide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
T80489-100G T80489-25G
SIALT80489-100GEA 102 EUR
SIALT80489-100G SIALT80489-25G
Trimethylsulphonium iodide, Sigma-Aldrich®
Trimethylsulphonium iodide
Formula: C₃H₉IS
MW: 204.07 g/mol
Melting Pt: 215…220 °C
Density: 1.922 g/cm³ (20 °C)
MDL Number: MFCD00011632
CAS Number: 2181-42-2
EINECS: 218-555-4