Uracil, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Pyrimidine-2,4(1H,3H)-dione
M-Clarity™ quality level = MQ200
U1128-100G
U1128-25G
SIALU1128-100GEA
184
EUR
SIALU1128-100G
SIALU1128-25G
Uracil, Sigma-Aldrich®
Uracil
Formula:
C₄H₄N₂O₂ MW: 112.09 g/mol Boiling Pt: 379 °C (1013 hPa) Melting Pt: 194 °C Density: 0.99744 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006016 CAS Number: 66-22-8 EINECS: 200-621-9 Merck Index: 12,09985 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...