Benzyl formate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Formic acid benzyl ester
M-Clarity™ quality level = MQ300
W214507-10KG-K
W214507-1KG-K
W214507-5KG-K
SIALW214507-10KG-KEA
333
EUR
SIALW214507-10KG-K
SIALW214507-1KG-K
SIALW214507-5KG-K
Benzyl formate, Sigma-Aldrich®
Benzyl formate
Formula:
C₈H₈O₂ MW: 136.15 g/mol |
MDL Number:
MFCD00014128 CAS Number: 104-57-4 EINECS: 203-214-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...