Cinnamaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
M-Clarity™ quality level = MQ300
W228613-100G-K W228613-1KG-K W228613-5KG-K W228613-SAMPLE
SIALW228613-100G-KEA 92.7 EUR
SIALW228613-100G-K SIALW228613-1KG-K SIALW228613-5KG-K SIALW228613-SAMPLE
Cinnamaldehyde, Sigma-Aldrich®
Cinnamaldehyde
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.112 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00007000
CAS Number: 104-55-2
EINECS: 203-213-9
Merck Index: 12,02356