Ethyl-10-undecenoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 10-Undecenoic acid ethyl ester
M-Clarity™ quality level = MQ300
W246107-100G W246107-1KG W246107-SAMPLE
SIALW246107-100GEA 83 EUR
SIALW246107-100G SIALW246107-1KG SIALW246107-SAMPLE
Ethyl-10-undecenoate, Sigma-Aldrich®
Ethyl 10-undecenoate
Formula: C₁₃H₂₄O₂
MW: 212.33 g/mol
Boiling Pt: 258 °C (1013 hPa)
Density: 0.879 g/cm³ (25 °C)
MDL Number: MFCD00009220
CAS Number: 692-86-4
EINECS: 211-734-8