4-Hydroxybenzylacetone, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W258814-1KG-K W258814-25G-K
SIALW258814-1KG-KEA 962 EUR
SIALW258814-1KG-K SIALW258814-25G-K
4-Hydroxybenzylacetone, Sigma-Aldrich®
4-Hydroxybenzylacetone
Formula: C₁₀H₁₂O₂
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 83…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00002394
CAS Number: 5471-51-2
EINECS: 226-806-4