Levulinic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid
M-Clarity™ quality level = MQ300
W262706-10KG-K
W262706-1KG-K
W262706-25KG-K
W262706-5KG-K
SIALW262706-10KG-KEA
324
EUR
SIALW262706-10KG-K
SIALW262706-1KG-K
SIALW262706-25KG-K
SIALW262706-5KG-K
Levulinic acid, Sigma-Aldrich®
Levulinic acid
Formula:
C₅H₈O₃ MW: 116.12 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 26…29 °C Density: 1.14 g/cm³ (20 °C) Flash Pt: 138 °C |
MDL Number:
MFCD00002796 CAS Number: 123-76-2 EINECS: 204-649-2 UN: 3261 ADR: 8,III Merck Index: 13,05492 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...