3-(Methylthio)propanal, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-(Methylthio)propionaldehyde

M-Clarity™ quality level = MQ300

W274704-1KG-K W274704-25KG-K W274704-5KG-K
SIALW274704-1KG-KEA 261 EUR
SIALW274704-1KG-K SIALW274704-25KG-K SIALW274704-5KG-K
3-(Methylthio)propanal, Sigma-Aldrich®
3-(Methylthio)propanal
Formula: C₄H₈OS
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –75 °C
Density: 1.036 g/cm³ (20 °C)
MDL Number: MFCD00007022
CAS Number: 3268-49-3
EINECS: 221-882-5
UN: 2785
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR