Disodium succinate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,4-Butanedioic acid disodium salt, Sodium Succinate, Succinic acid disodium salt, Sodium succinate dibasic
M-Clarity™ quality level = MQ300
W327700-10KG-K W327700-1KG-K W327700-5KG-K
SIALW327700-10KG-KEA 407 EUR
SIALW327700-10KG-K SIALW327700-1KG-K SIALW327700-5KG-K
Disodium succinate, Sigma-Aldrich®
Disodium succinate
Formula: C₄H₄Na₂O₄
MW: 162.05 g/mol
Density: 1.88 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002790
CAS Number: 150-90-3
EINECS: 205-778-7
Merck Index: 12,08828