2-Isopropyl-4-methylthiazole, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W355518-100G-K W355518-1KG-K W355518-5KG-K
SIALW355518-100G-KEA 208 EUR
SIALW355518-100G-K SIALW355518-1KG-K SIALW355518-5KG-K
2-Isopropyl-4-methylthiazole, Sigma-Aldrich®
2-Isopropyl-4-methylthiazole
Formula: C₇H₁₁NS
MW: 141.24 g/mol
MDL Number: MFCD00036561
CAS Number: 15679-13-7
EINECS: 239-758-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR