Thiazole, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W361518-100G-K W361518-1KG-K W361518-25G-K
SIALW361518-100G-KEA 581 EUR
SIALW361518-100G-K SIALW361518-1KG-K SIALW361518-25G-K
Thiazole, Sigma-Aldrich®
Thiazole
Formula: C₃H₃NS
MW: 85.13 g/mol
Boiling Pt: 116.8 °C (1013 hPa)
Melting Pt: –34 °C
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00005315
CAS Number: 288-47-1
EINECS: 206-021-3
Merck Index: 12,09442

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR