Methyl methacrylate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methacrylic acid methyl ester
M-Clarity™ quality level = MQ300
W400201-10KG W400201-1KG W400201-5KG W400201-SAMPLE
SIALW400201-10KGEA 495 EUR
SIALW400201-10KG SIALW400201-1KG SIALW400201-5KG SIALW400201-SAMPLE
Methyl methacrylate, Sigma-Aldrich®
Methyl methacrylate (monomer)
Formula: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008587
CAS Number: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3,II
REACH: 01-2119452498-28
Merck Index: 12,06005