Picric acid, yellow, fluid
Supplier: PanReac AppliChem
Synonyms: 2,4,6-Trinitrophenol, TNP
251049.1610 251049.1612
APPC251049.1610EA 88.6 EUR
APPC251049.1610 APPC251049.1612
Picric acid, yellow, fluid
Picric acid
Formula: C₆H₃N₃O₇
MW: 229.11 g/mol
Boiling Pt: 100 °C (212 °F)
Melting Pt: 80.35
Density: 1,006 kg/L
Flash Pt: 150
Storage Temperature: Room Temperature
MDL Number: MFCD00007102
CAS Number: 88-89-1
Merck Index: 12,07562