ESD Safety Shoes, Low, MATTIS XXSports, S1P
Supplier: Elten
ELTN720321/44EA 90.9 EUR
ELTN720321/44 ELTN720321/43 ELTN720321/46 ELTN720321/35 ELTN720321/45 ELTN720321/37 ELTN720321/48 ELTN720321/36 ELTN720321/47 ELTN720321/39 ELTN720321/38 ELTN720321/40 ELTN720321/42 ELTN720321/41
ESD Safety Shoes, Low, MATTIS XXSports, S1P
Shoes and Boots
Textile material with recycled material content.
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD black
  • Non-metallic, penetration-resistant midsolewith recycled material content
  • TPU/PU sole WELLMAXX SPORTS
  • Steel toe cap
Designed for use in dry industry, storage, transport, and assembly work areas where there is a risk of penetration from pointed and sharp objects (S1P). Shoes are also suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD) and workplaces on hard undergrounds. The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides, creating more energy in every step.
Certifikace: EN ISO 20345 S1P SRB, form A.