(R)-(+)-Citronellal, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(R)-(+)-3,7-Dimethyloct-6-enal
M-Clarity™ quality level = MQ100
343641-1G
343641-5G
SIAL343641-1GEA
131
EUR
SIAL343641-1G
SIAL343641-5G
(R)-(+)-Citronellal, Sigma-Aldrich®
(R)-(+)-Citronellal
Formula:
C₁₀H₁₈O MW: 154.25 g/mol |
CAS Number:
2385-77-5 EINECS: 219-194-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...