(S)-(-)-Citronellal, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(-)-3,7-Dimethyl-6-octenal, (S)-(-)-3,7-Dimethyloct-6-enal, (-)-Citronellal, (S)-3,7-Dimethyloct-6-enal
M-Clarity™ quality level = MQ100
373753-1G 373753-5G
SIAL373753-1GEA 47.5 EUR
SIAL373753-1G SIAL373753-5G
(S)-(-)-Citronellal, Sigma-Aldrich®
(S)-(-)-Citronellal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 84 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.85 g/cm³ (20 °C)
MDL Number: MFCD00075588
CAS Number: 5949-05-3
EINECS: 227-707-9